Triphenylmethane

Supplier: Apollo Scientific
Warning

OR0040-100G OR0040-1KG OR0040-5KG
APOSOR0040-100GEA 21.9 EUR
APOSOR0040-100G APOSOR0040-1KG APOSOR0040-5KG
Triphenylmethane
Triphenylmethane
Formula: C₁₉H₁₆
Boiling Pt: 358…359 °C/754mmHg
Melting Pt: 92…94 °C
Density: 1,014 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004763
CAS Number: 519-73-3
EINECS: 208-275-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR