1-Hexyne
Supplier: Apollo Scientific
|
Danger
|
OR0049-250G
OR0049-50G
APOSOR0049-250GEA
401
EUR
APOSOR0049-250G
APOSOR0049-50G
1-Hexyne
1-Hexyne
Formula:
C₆H₁₀ MW: 82,15 g/mol Boiling Pt: 71…72 °C Melting Pt: –132 °C Density: 0,715 g/cm³ (25 °C) Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009504 CAS Number: 693-02-7 EINECS: 211-736-9 UN: 3295 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...