2,3-Dihydroxybenzoic acid

Supplier: Apollo Scientific
Warning

Synonyms: Pyrocatechuic acid

OR0057-100G OR0057-25G OR0057-5G
APOSOR0057-100GEA 404 EUR
APOSOR0057-100G APOSOR0057-25G APOSOR0057-5G
2,3-Dihydroxybenzoic acid
2,3-Dihydroxybenzoic acid
Formula: C₇H₆O₄
MW: 154,12 g/mol
Boiling Pt: 95…96 °C/0,5mmHg
Melting Pt: 208…210 °C
Storage Temperature: Ambient
MDL Number: MFCD00002446
CAS Number: 303-38-8
EINECS: 206-139-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR