3,5-Dichlorophenol

Supplier: Apollo Scientific
Danger

OR0059-100G OR0059-25G OR0059-5G
APOSOR0059-100GEA 89.9 EUR
APOSOR0059-100G APOSOR0059-25G APOSOR0059-5G
3,5-Dichlorophenol
3,5-Dichlorophenol
Formula: C₆H₄Cl₂O
Boiling Pt: 233 °C
Melting Pt: 63…68 °C
Storage Temperature: Ambient
MDL Number: MFCD00002259
CAS Number: 591-35-5
EINECS: 209-714-9
UN: 2020
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR