Ethyl-4-piperidinecarboxylate 97%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
Ethyl piperidine-4-carboxylate, Ethyl isonipecotate, Ethyl 4-Piperidinecarboxylate
OR0070-500G
APOSOR0070-500GEA
92.8
EUR
APOSOR0070-500G
Ethyl-4-piperidinecarboxylate 97%
Ethyl 4-piperidinecarboxylate
Formula:
C₈H₁₅NO₂ MW: 157,21 g/mol Boiling Pt: 204 °C Density: 1,02 g/cm³ (25 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006003 CAS Number: 1126-09-6 EINECS: 214-416-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...