Trimellitic acid 98%

Supplier: Apollo Scientific

Synonyms: 1,2,4-Benzenetricarboxylic acid, Benzene-1,2,4-tricarboxylic acid

OR0075-100G OR0075-25G OR0075-500G
APOSOR0075-100GEA 92.8 EUR
APOSOR0075-100G APOSOR0075-25G APOSOR0075-500G
Trimellitic acid 98%
Trimellitic acid
Formula: C₉H₆O₆
MW: 210.14 g/mol
Melting Pt: 231 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002470
CAS Number: 528-44-9
Merck Index: 12,09832

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR