Sulphamethoxazole

Supplier: Apollo Scientific
Warning

Synonyms: 4-Amino-N-(5-methylisoxazol-3-yl)benzenesulphonamide

OR0144-10G OR0144-5G
APOSOR0144-10GEA 21.9 EUR
APOSOR0144-10G APOSOR0144-5G
Sulphamethoxazole
Sulphamethoxazole
Formula: C₁₀H₁₁N₃O₃S
MW: 253,28 g/mol
Melting Pt: 166…169 °C
Storage Temperature: Ambient
MDL Number: MFCD00010546
CAS Number: 723-46-6
EINECS: 211-963-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR