3-Hydroxypyridine-2-carboxaldehyde
Supplier: Apollo Scientific
|
Warning
|
OR0151-5G
OR0151-1G
OR0151-10G
APOSOR0151-5GEA
463
EUR
APOSOR0151-5G
APOSOR0151-1G
APOSOR0151-10G
3-Hydroxypyridine-2-carboxaldehyde
3-Hydroxypyridine-2-carboxaldehyde
Formula:
C₆H₅NO₂ Boiling Pt: 96…100 °C/30mmHg Melting Pt: 80…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD06245505 CAS Number: 1849-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...