Ethyl bromopyruvate, Technical Grade
Supplier: Apollo Scientific
|
Danger
|
OR015293-100G
OR015293-250G
OR015293-25G
APOSOR015293-100GEA
43.5
EUR
APOSOR015293-100G
APOSOR015293-250G
APOSOR015293-25G
Ethyl bromopyruvate, Technical Grade
Ethyl bromopyruvate
Formula:
BrH₂CC(O)COOCH₂CH₃ Boiling Pt: 98…100 °C/10mmHg Density: 1,554 g/cm³ (25 °C) Flash Pt: 99 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00000204 CAS Number: 70-23-5 EINECS: 200-729-6 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...