2-Nitrophenol

Supplier: Apollo Scientific
Danger

OR01570-25G OR01570-5G
APOSOR01570-25GEA 46.4 EUR
APOSOR01570-25G APOSOR01570-5G
2-Nitrophenol
2-Nitrophenol
Formula: C₆H₅NO₃
Boiling Pt: 214…216 °C
Melting Pt: 44…48 °C
Density: 1,26 g/cm³ (25 °C)
Flash Pt: 108 °C
Storage Temperature: Ambient
MDL Number: MFCD00011688
CAS Number: 88-75-5
UN: 1663
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR