5-Methyl-4H-1,2,4-triazol-3-amine 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Amino-5-methyl-4H-1,2,4-triazole
OR01571-1G
OR01571-5G
APOSOR01571-1GEA
52.2
EUR
APOSOR01571-1G
APOSOR01571-5G
5-Methyl-4H-1,2,4-triazol-3-amine 98%
5-Methyl-4H-1,2,4-triazol-3-amine
Formula:
C3H6N4 MW: 98,1100 g/mol Melting Pt: 142…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD00498236 CAS Number: 4923-01-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...