4-(1-Hydroxyethyl)pyridine
Supplier: Apollo Scientific
Synonyms:
1-(Pyridin-4-yl)ethanol, 1-(Pyridin-4-yl)ethan-1-ol
OR01620-1G
OR01620-5G
OR01620-25G
APOSOR01620-1GEA
68.3
EUR
APOSOR01620-1G
APOSOR01620-5G
APOSOR01620-25G
4-(1-Hydroxyethyl)pyridine
4-(1-Hydroxyethyl)pyridine
Formula:
C₇H₉NO MW: 123,15 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00077867 CAS Number: 23389-75-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...