4-Benzoyl benzylamine hydrochloride 95+%

Supplier: Apollo Scientific
Warning

OR01642-1G OR01642-5G OR01642-250MG
APOSOR01642-1GEA 509 EUR
APOSOR01642-1G APOSOR01642-5G APOSOR01642-250MG
4-Benzoyl benzylamine hydrochloride 95+%
4-Benzoyl benzylamine hydrochloride
Formula: C₁₄H₁₄ClNO
MW: 247,73 g/mol
Melting Pt: 207…208 °C
Storage Temperature: Ambient
MDL Number: MFCD06245504
CAS Number: 24095-40-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR