tert-Butyl-4-aminophenylacetate 95+%

Supplier: Apollo Scientific
Warning

Synonyms: tert-Butyl 2-(4-aminophenyl)acetate

OR01647-5G
APOSOR01647-5GEA 140 EUR
APOSOR01647-5G
tert-Butyl-4-aminophenylacetate 95+%
tert-Butyl-4-aminophenylacetate
Formula: C₁₂H₁₇NO₂
MW: 207,27 g/mol
Melting Pt: 30…32 °C
Storage Temperature: Ambient
MDL Number: MFCD06245494
CAS Number: 174579-31-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR