3-Bromoisonicotinic acid 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Bromopyridine-4-carboxylic acid, 3-Bromo-4-pyridinecarboxylic acid
OR0167-25G
OR0167-5G
OR0167-100G
APOSOR0167-25GEA
45.2
EUR
APOSOR0167-25G
APOSOR0167-5G
APOSOR0167-100G
3-Bromoisonicotinic acid 98%
3-Bromoisonicotinic acid
Formula:
C₆H₄BrNO₂ MW: 202,01 g/mol Melting Pt: 234…238 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040944 CAS Number: 13959-02-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...