(4-Bromophenyl)acetonitrile 97%

Supplier: Apollo Scientific
Danger

Synonyms: 4-Bromophenylacetonitrile, 4-Bromobenzyl cyanide

OR017366-1G OR017366-5G OR017366-10G
APOSOR017366-1GEA 21.9 EUR
APOSOR017366-1G APOSOR017366-5G APOSOR017366-10G
(4-Bromophenyl)acetonitrile 97%
(4-Bromophenyl)acetonitrile
Formula: C₈H₆BrN
MW: 196,05 g/mol
Boiling Pt: 286 °C
Melting Pt: 45…49 °C
Density: 1,49 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00001916
CAS Number: 16532-79-9
EINECS: 240-602-2
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR