ω-Bromo-4-methylacetophenone

Supplier: Apollo Scientific
Warning

Synonyms: α-Bromo-4-methylacetophenone, Bromomethyl p-methylphenyl ketone, 2-Bromo-4'-methylacetophenone, 2-Bromo-1-(p-tolyl)ethanone, 4-Methylphenacyl bromide

OR017854-1G OR017854-5G
APOSOR017854-1GEA 14.7 EUR
APOSOR017854-1G APOSOR017854-5G
ω-Bromo-4-methylacetophenone
ω-Bromo-4-methylacetophenone
Formula: C₉H₉BrO
MW: 213,07 g/mol
Boiling Pt: 105 °C/0,1mmHg
Melting Pt: 48…52 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000203
CAS Number: 619-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR