2-(1-Pyrrolyl)benzylamine 95+%

Supplier: Apollo Scientific
Danger

OR0180-1G OR0180-5G
APOSOR0180-1GEA 207 EUR
APOSOR0180-1G APOSOR0180-5G
2-(1-Pyrrolyl)benzylamine 95+%
2-(1-Pyrrolyl)benzylamine
Formula: C₁₁H₁₂N₂
MW: 172,23 g/mol
Boiling Pt: 80…82 °C/0,15mmHg
Storage Temperature: Ambient
MDL Number: MFCD03763920
CAS Number: 39243-88-4
UN: 2735
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR