1-(Bromomethyl)adamantane 95+%
Supplier: Apollo Scientific
|
Danger
|
OR018152-1G
OR018152-10G
OR018152-5G
APOSOR018152-1GEA
31.9
EUR
APOSOR018152-1G
APOSOR018152-10G
APOSOR018152-5G
1-(Bromomethyl)adamantane 95+%
1-(Bromomethyl)adamantane
Formula:
C₁₁H₁₇Br Melting Pt: 30…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00185303 CAS Number: 14651-42-4 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...