Ethyl-1,2,4-oxadiazole-3-carboxylate 95%

Supplier: Apollo Scientific
Warning

Synonyms: Ethyl 1,2,4-oxadiazole-3-carboxylate

OR0191-1G
APOSOR0191-1GEA 144 EUR
APOSOR0191-1G
Ethyl-1,2,4-oxadiazole-3-carboxylate 95%
Ethyl-1,2,4-oxadiazole-3-carboxylate
Formula: C₅H₆N₂O₃
MW: 142,12 g/mol
Boiling Pt: 88…90 °C/2,25mmHg
Melting Pt: 41…43 °C
Storage Temperature: Ambient
MDL Number: MFCD06245497
CAS Number: 39512-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR