5-Chloro-6-(4-chlorophenyl)pyridazin-3(2H)-one 95+%

Supplier: Apollo Scientific
Warning

OR0197-1G OR0197-5G OR0197-25G
APOSOR0197-1GEA 229 EUR
APOSOR0197-1G APOSOR0197-5G APOSOR0197-25G
5-Chloro-6-(4-chlorophenyl)pyridazin-3(2H)-one 95+%
5-Chloro-6-(4-chlorophenyl)pyridazin-3(2H)-one
Formula: C₁₀H₆Cl₂N₂O
MW: 241.08 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD06245502
CAS Number: 80591-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR