4-Aminobutyric acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
GABA, γ-Aminobutyric acid
OR01972-100G
OR01972-500G
APOSOR01972-100GEA
89.9
EUR
APOSOR01972-100G
APOSOR01972-500G
4-Aminobutyric acid
4-Aminobutyric acid
Formula:
C₄H₉NO₂ MW: 103.12 g/mol Boiling Pt: 248 Melting Pt: 195 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008226 CAS Number: 56-12-2 EINECS: 200-258-6 Merck Index: 13,00429 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...