4-Chloro-2-isocyanato-1-methoxybenzene 97%

Supplier: Apollo Scientific
Danger

Synonyms: 5-Chloro-2-methoxyphenyl isocyanate

OR0249-1G OR0249-5G
APOSOR0249-1GEA 34.8 EUR
APOSOR0249-1G APOSOR0249-5G
4-Chloro-2-isocyanato-1-methoxybenzene 97%
4-Chloro-2-isocyanato-1-methoxybenzene

When melted, may contain small amount of sediment.

Formula: C₈H₆ClNO₂
MW: 183,6 g/mol
Melting Pt: 58…62 °C
Storage Temperature: Ambient
MDL Number: MFCD00013860
CAS Number: 55440-54-5
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR