4-Chloro-o-toluic acid
Supplier: Apollo Scientific
|
Danger
|
OR0251-100G
OR0251-25G
OR0251-5G
APOSOR0251-100GEA
217
EUR
APOSOR0251-100G
APOSOR0251-25G
APOSOR0251-5G
4-Chloro-o-toluic acid
4-Chloro-o-toluic acid
Formula:
C₈H₇ClO₂ MW: 170,6 g/mol Melting Pt: 167…174 °C Storage Temperature: Ambient |
MDL Number:
MFCD00045852 CAS Number: 7499-07-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...