4-Chloro-3-nitrophenyl isocyanate
Supplier: Apollo Scientific
|
Danger
|
OR0268-5G
OR0268-1G
APOSOR0268-5GEA
75.7
EUR
APOSOR0268-5G
APOSOR0268-1G
4-Chloro-3-nitrophenyl isocyanate
4-Chloro-3-nitrophenyl isocyanate
When melted, may contain small amount of sediment.
Formula:
C₇H₃ClN₂O₃ MW: 198,57 g/mol Melting Pt: 63…69 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00037062 CAS Number: 40397-96-4 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...