ω-Bromo-2-chloroacetophenone 95%

Supplier: Apollo Scientific
Danger

Synonyms: α-Bromo-2-chloroacetophenone, Bromomethyl o-chlorophenyl ketone, 2-Bromo-1-(2-chlorophenyl)ethanone, 2-Bromo-2'-chloroacetophenone, 2-Chlorophenacyl bromide

OR0270-25G
APOSOR0270-25GEA 78.3 EUR
APOSOR0270-25G
ω-Bromo-2-chloroacetophenone 95%
ω-Bromo-2-chloroacetophenone
Formula: C₈H₆BrClO
MW: 233,49 g/mol
Boiling Pt: 162…166 °C/20mmHg
Density: 1,602 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00832993
CAS Number: 5000-66-8
UN: 2922
ADR: 8+6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR