4-[(4-Chlorophenyl)sulphanyl]-3-nitrobenzenecarbaldehyde
Supplier: Apollo Scientific
Synonyms:
4-(4-Chlorophenylthio)-3-nitrobenzaldehyde
OR0292-1G
OR0292-250MG
APOSOR0292-1GEA
0
EUR
APOSOR0292-1G
APOSOR0292-250MG
4-[(4-Chlorophenyl)sulphanyl]-3-nitrobenzenecarbaldehyde
4-[(4-Chlorophenyl)sulphanyl]-3-nitrobenzenecarbaldehyde
Formula:
C₁₃H₈ClNO₃S MW: 293,73 g/mol Melting Pt: 120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00098773 CAS Number: 270262-96-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...