2,3-Dichlorobenzoyl cyanide
Supplier: Apollo Scientific
Synonyms:
(2,3-Dichlorophenyl)(oxo)acetonitrile
OR0344-100G
OR0344-25G
APOSOR0344-100GEA
207
EUR
APOSOR0344-100G
APOSOR0344-25G
2,3-Dichlorobenzoyl cyanide
2,3-Dichlorobenzoyl cyanide
Formula:
C₈H₃Cl₂NO MW: 200,03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00792559 CAS Number: 77668-42-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...