1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid
Supplier: Apollo Scientific
|
Warning
|
OR0563-1G
OR0563-250MG
OR0563-5G
APOSOR0563-1GEA
88.6
EUR
APOSOR0563-1G
APOSOR0563-250MG
APOSOR0563-5G
1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid
1-Isopropyl-1H-1,2,3-benzotriazole-5-carboxylic acid
Formula:
C₁₀H₁₁N₃O₂ MW: 205,22 g/mol Melting Pt: 222…231 °C Storage Temperature: Ambient |
MDL Number:
MFCD01570664 CAS Number: 306935-41-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...