6-Methoxyindole 98%

Supplier: Apollo Scientific
Warning

Synonyms: 6-Methoxy-1H-indole, 1H-Indol-6-yl methyl ether, Indol-6-yl methyl ether

OR0581-1G OR0581-25G OR0581-10G
APOSOR0581-1GEA 21.9 EUR
APOSOR0581-1G APOSOR0581-25G APOSOR0581-10G
6-Methoxyindole 98%
6-Methoxyindole
Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 105 °C/0.2mmHg
Melting Pt: 90…94 °C
Storage Temperature: Ambient
MDL Number: MFCD00022780
CAS Number: 3189-13-7
EINECS: 221-689-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR