3-Methylbenzamidoxime
Supplier: Apollo Scientific
|
Warning
|
OR0604-1G
OR0604-5G
APOSOR0604-1GEA
45.1
EUR
APOSOR0604-1G
APOSOR0604-5G
3-Methylbenzamidoxime
3-Methylbenzamidoxime
Formula:
C₈H₁₀N₂O Melting Pt: 87…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00833266 CAS Number: 40067-82-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...