2-(2-Naphthyl)indole 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
2-(Naphthalen-2-yl)-1H-indole
OR0668-1G
OR0668-5G
OR0668-250MG
APOSOR0668-1GEA
143
EUR
APOSOR0668-1G
APOSOR0668-5G
APOSOR0668-250MG
2-(2-Naphthyl)indole 98%
2-(2-Naphthyl)indole
Formula:
C₁₈H₁₃N Melting Pt: 204…207 °C Storage Temperature: Ambient |
MDL Number:
MFCD00124144 CAS Number: 23746-81-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...