4-Nitro-L-phenylalanine
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
H-Phe(4-NO2)-OH, (S)-4-Nitrophenylalanine, (S)-2-Amino-3-(4-nitrophenyl)propanoic acid
OR0679-25G
OR0679-100G
APOSOR0679-25GEA
21.9
EUR
APOSOR0679-25G
APOSOR0679-100G
4-Nitro-L-phenylalanine
4-Nitro-L-phenylalanine
Formula:
C₉H₁₀N₂O₄ Melting Pt: 221,9…223.3 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051221 CAS Number: 949-99-5 EINECS: 213-446-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...