4-(Methylthio)aniline 97%

Supplier: Apollo Scientific
Warning

Synonyms: 4-(Methylmercapto)aniline

OR0773-100G OR0773-25G OR0773-5G
APOSOR0773-100GEA 215 EUR
APOSOR0773-100G APOSOR0773-25G APOSOR0773-5G
4-(Methylthio)aniline 97%
4-(Methylthio)aniline
Formula: C₇H₉NS
MW: 139,22 g/mol
Boiling Pt: 272…273 °C
Density: 1,119 g/cm³ (25 °C)
Flash Pt: 110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007889
CAS Number: 104-96-1
EINECS: 203-256-3
UN: 3334
ADR: 9,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR