4-Iodo-2,6-xylenol

Warning

OR0805-10G OR0805-1G OR0805-25G
APOSOR0805-10GEA 71.3 EUR
APOSOR0805-10G APOSOR0805-1G APOSOR0805-25G
4-Iodo-2,6-xylenol
4-Iodo-2,6-xylenol
Formula: C₈H₉IO
MW: 248,06 g/mol
Melting Pt: 96…101 °C
Storage Temperature: Ambient
MDL Number: MFCD00094415
CAS Number: 10570-67-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR