2-Phenylpiperidine
Supplier: Apollo Scientific
|
Warning
|
OR0819-100MG
OR0819-1G
APOSOR0819-100MGEA
21.9
EUR
APOSOR0819-100MG
APOSOR0819-1G
2-Phenylpiperidine
2-Phenylpiperidine
Formula:
C₁₁H₁₅N Boiling Pt: 70…72 °C/0,45mmHg Storage Temperature: Ambient |
MDL Number:
MFCD00270125 CAS Number: 3466-80-6 EINECS: 222-422-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...