2-Chlorobenzophenone 98%
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
o-Chlorobenzophenone, (2-Chlorophenyl)(phenyl)methanone
OR0833-100G
OR0833-500G
OR0833-1KG
APOSOR0833-100GEA
21.9
EUR
APOSOR0833-100G
APOSOR0833-500G
APOSOR0833-1KG
2-Chlorobenzophenone 98%
2-Chlorobenzophenone
Formula:
C₁₃H₉ClO MW: 216,67 g/mol Boiling Pt: 180 °C/15mmHg Melting Pt: 45…47 °C Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000558 CAS Number: 5162-03-8 EINECS: 225-936-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...