1,2,3-Trichloro-5-nitrobenzene

Supplier: Apollo Scientific
Warning

OR0843-25G OR0843-5G
APOSOR0843-25GEA 60.9 EUR
APOSOR0843-25G APOSOR0843-5G
1,2,3-Trichloro-5-nitrobenzene
1,2,3-Trichloro-5-nitrobenzene
Formula: C₆H₂Cl₃NO₂
Boiling Pt: 247 °C
Melting Pt: 68…71 °C
Density: 1,126 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00061096
CAS Number: 20098-48-0
EINECS: 243-511-6
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR