Hydroxylamine-O-sulphonic acid 98%

Supplier: Apollo Scientific
Danger

Synonyms: Amino hydrogen sulphate

OR0911-100G OR0911-250G OR0911-1KG
APOSOR0911-100GEA 51 EUR
APOSOR0911-100G APOSOR0911-250G APOSOR0911-1KG
Hydroxylamine-O-sulphonic acid 98%
Hydroxylamine-O-sulphonic acid
Formula: H₃NO₄S
MW: 113,09 g/mol
Melting Pt: 208…211 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011604
CAS Number: 2950-43-8
EINECS: 220-971-6
UN: 3260
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR