Isonicotinamide 99%

Supplier: Apollo Scientific
Warning

OR0914-100G OR0914-25G
APOSOR0914-100GEA 58 EUR
APOSOR0914-100G APOSOR0914-25G
Isonicotinamide 99%
Isonicotinamide
Formula: C₆H₆N₂O
Melting Pt: 155…157 °C
Storage Temperature: Ambient
MDL Number: MFCD00006432
CAS Number: 1453-82-3
EINECS: 215-926-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR