3-Piperazin-1-ylpropionic acid 98%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(1-Piperazinyl)propionic acid, 3-(Piperazin-1-yl)propanoic acid

OR0947-1G OR0947-250MG OR0947-2G
APOSOR0947-1GEA 60.9 EUR
APOSOR0947-1G APOSOR0947-250MG APOSOR0947-2G
3-Piperazin-1-ylpropionic acid 98%
3-Piperazin-1-ylpropionic acid
Formula: C₇H₁₄N₂O₂
MW: 158,2 g/mol
Melting Pt: 199…201 °C
Storage Temperature: Ambient
MDL Number: MFCD03161179
CAS Number: 27245-31-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR