3-Azetidinecarboxylic acid 96%

Supplier: Apollo Scientific
Warning

Synonyms: 3-Azetanecarboxylic acid, Azetidine-3-carboxylic acid

OR0961-5G OR0961-25G OR0961-100G
APOSOR0961-5GEA 21.9 EUR
APOSOR0961-5G APOSOR0961-25G APOSOR0961-100G
3-Azetidinecarboxylic acid 96%
3-Azetidinecarboxylic acid
Formula: C₄H₇NO₂
MW: 101,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00191763
CAS Number: 36476-78-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR