Cyclopropanecarbonitrile 94%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Cyclopropyl cyanide
OR0999-100G
OR0999-25G
APOSOR0999-100GEA
139
EUR
APOSOR0999-100G
APOSOR0999-25G
Cyclopropanecarbonitrile 94%
Cyclopropanecarbonitrile
Formula:
C₄H₅N MW: 67,09 g/mol Boiling Pt: 133…135 °C Density: 0,905 g/cm³ (25 °C) Flash Pt: 32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00001269 CAS Number: 5500-21-0 EINECS: 226-836-8 UN: 3275 ADR: 6.1+3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...