3-Amino-6-chloro-4,5-dimethylpyridazine 95+%

Supplier: Apollo Scientific
Warning

OR1003-1G OR1003-5G
APOSOR1003-1GEA 81.2 EUR
APOSOR1003-1G APOSOR1003-5G
3-Amino-6-chloro-4,5-dimethylpyridazine 95+%
3-Amino-6-chloro-4,5-dimethylpyridazine
Formula: C₆H₈ClN₃
Melting Pt: 202…206 °C
Storage Temperature: Ambient
MDL Number: MFCD00828815
CAS Number: 76593-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR