3-Phenylpropionic acid
Supplier: Apollo Scientific
|
Warning
|
Synonyms:
3-Phenylpropanoic acid, Hydrocinnamic acid
OR10046-100G
OR10046-500G
APOSOR10046-100GEA
35
EUR
APOSOR10046-100G
APOSOR10046-500G
3-Phenylpropionic acid
3-Phenylpropionic acid
Formula:
C₉H₁₀O₂ MW: 150,18 g/mol Boiling Pt: 280 °C Melting Pt: 45…48 °C Density: 1,071 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00002771 CAS Number: 501-52-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...