1-Ethylpropylamine 99%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Pentylamine, 3-Aminopentane, Pentan-3-amine
OR10080-500G
APOSOR10080-500GEA
342
EUR
APOSOR10080-500G
1-Ethylpropylamine 99%
1-Ethylpropylamine
Formula:
C₅H₁₃N MW: 87,16 g/mol Boiling Pt: 88…91 °C Density: 0,748 g/cm³ (25 °C) Flash Pt: 2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008096 CAS Number: 616-24-0 UN: 2733 ADR: 3+8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...