Cyclopentanol 97%

Supplier: Apollo Scientific
Warning

OR10094-100G OR10094-1KG OR10094-500G
APOSOR10094-100GEA 94.4 EUR
APOSOR10094-100G APOSOR10094-1KG APOSOR10094-500G
Cyclopentanol 97%
Cyclopentanol
Formula: C₅H₁₀O
Boiling Pt: 139…141 °C
Melting Pt: –18 °C
Density: 0,95 g/cm³ (25 °C)
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00001363
CAS Number: 96-41-3
UN: 2244
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR