3-Aminothiophenol 97%
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
3-Aminophenyl mercaptan, 3-Mercaptoaniline, 3-Aminobenzenethiol
OR1010-100G
OR1010-25G
OR1010-5G
APOSOR1010-100GEA
214
EUR
APOSOR1010-100G
APOSOR1010-25G
APOSOR1010-5G
3-Aminothiophenol 97%
3-Aminothiophenol
Formula:
C₆H₇NS MW: 125.19 g/mol Boiling Pt: 123…125 °C/8mmHg Density: 1.179 g/cm³ (25 °C) Flash Pt: 79 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00007791 CAS Number: 22948-02-3 EINECS: 245-344-4 UN: 3267 ADR: 8 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...