Dioctylamine
Supplier: Apollo Scientific
|
Danger
|
Synonyms:
Di-n-octylamine, N-Octyl-1-octanamine
OR10117-100G
OR10117-25G
APOSOR10117-100GEA
132
EUR
APOSOR10117-100G
APOSOR10117-25G
Dioctylamine
Dioctylamine
Formula:
C₁₆H₃₅N MW: 241,46 g/mol Boiling Pt: 297…298 °C Melting Pt: 13…16 °C Density: 0,799 g/cm³ (25 °C) Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009557 CAS Number: 1120-48-5 EINECS: 214-311-6 UN: 2735 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...