3-(4-Nitrophenoxy)propionic acid 97+%

Supplier: Apollo Scientific
Warning

Synonyms: 3-(4-Nitrophenoxy)propanoic acid

OR10128-1G OR10128-5G
APOSOR10128-1GEA 39.3 EUR
APOSOR10128-1G APOSOR10128-5G
3-(4-Nitrophenoxy)propionic acid 97+%
3-(4-Nitrophenoxy)propionic acid
Formula: C₉H₉NO₅
MW: 211.1700 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00043748
CAS Number: 10572-16-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR